Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Total BDNF Quantikine ELISA Kit
SDP

Catalog No. SBNT00 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SBNT00 1 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. SBNT00 Supplier R&D Systems™ Supplier No. SBNT00
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Total BDNF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Total BDNF Quantikine ELISA Kit quantifies human, mouse, rat, porcine, canine BDNF in cell culture supernates (50 ul), tissue lysates (50 ul), serum (10 ul), edta plasma (50 ul), platelet-poor edta plasma (10 ul), heparin plasma (50 ul), platelet-poor heparin plasma (10 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 1.35pg/mL
Conjugate HRP
Cross Reactivity < 0.5% cross-reactivity observed with available related molecules, < 50% cross-species reactivity observed with species tested
Product Type Total BDNF Quantikine ELISA Kit
Sample Type Cell Culture Supernatant, Human Milk, Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Serum, Tissue Lysates, Urine
Specificity Natural and Recombinant free, trk-bound, pro-, and mature BDNF.
For Use With (Application) ELISA
Gene ID (Entrez)
  • 627 (Human)
  • 12064 (Mouse)
  • 24225 (Rat)
Precision
  • Intra-Assay Precision (Precision within an assay) Three samples of known concentration were tested twenty times on one plate to assess intra-assay precision
  • Inter-Assay Precision (Precision between assays) Three samples of known concentration were tested in twenty separate assays to assess inter-assay precision. Assays were performed by at least three technicians using two lots of components
Quantity 1 Pack
Sample Volume Cell Culture Supernates (50uL),Tissue Lysates (50uL),Serum (10uL),EDTA Plasma (50uL),Platelet-poor EDTA Plasma (10uL),Heparin Plasma (50uL),Platelet-poor Heparin Plasma (10uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.