Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pointe Scientific Total Bilirubin Reagents

Catalog No. 23666151 Shop All Pointe Scientific Products
Change view
Click to view available options
Quantity:
1 x 120 mL
1 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-151 1 x 1 L
23-666-150 1 x 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-666-151 Supplier Pointe Scientific Supplier No. B75761L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Total bilirubin reagent: Sulfanilic acid 16mM, hydrochloric acid 164mM, dimethyl sulfoxide 4.4M, surfactant; Sodium nitrite reagent: Sodium nitrite 0.6M

For quantifying total bilirubin

  • Method: Walters and Gerarde, Endpoint
  • Format: Liquid (ready to use)
  • Wavelength: 550nm
  • Linearity: 20mg/dL
  • Expected Values: 0.2 to 1.2mg/dL

For the quantitative determination of total bilirubin in serum.

Specifications

Certifications/Compliance CE Marked
Type Total Bilirubin Reagent Kit
Formulation Total Bilirubin reagent: Sulfanilic Acid 16mM, Hydrochloric Acid 164mM, Dimethyl Sulfoxide 4.4M, Surfactants, Sodium Nitrite 0.6M
For Use With (Application) Quantitative determination of total Bilirubin in Serum, in-vitro diagnostic use only
Quantity 1 x 1 L
Wavelength 550 nm
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.