Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedical™ Total Estrogens Double Antibody RIA Kit

Catalog No. MP07140202 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 Tubes
500 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP07140202 100 Tubes
MP07140205 500 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP07140202 Supplier MP Biomedical™ Supplier No. 07140202
Add to Cart
Add to Cart

Quantitative Liquid Phase Radioimmunoassay

  • Total Estrogens Double Antibody RIA Kit detects the combined concentrations of unconjugated estradiol (E2) and estrone (E1).
  • Does not detect estrogen sulfates or glucuronides
  • Will not distinguish between free E1 or E2 or E2 bound to circulating binding proteins
  • Total Estrogen measurements are an approximation of plasma estrogenicity since E2 is biologically more active than E1 and the E2/E1 ratio may fluctuate widely under varying physiological and pathological conditions.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Kit and Assay
Sample Type Plasma, Serum
Target Species Human
For Use With (Application) Endocrinology
Quantity 100 Tubes
Sample Volume 500 μL
Storage Requirements 2°C to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.