Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Total MMP-2 Quantikine ELISA Kit
GSA_VA

Catalog No. MMP200
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMP200 1 Plate
SMMP200 1 Pk. (6 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MMP200 Supplier R&D Systems™ Supplier No. MMP200
Add to Cart
Add to Cart
This item is not returnable. View return policy

For human, mouse, rat, porcine, and canine samples.

The Total MMP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat, porcine, canine MMP-2 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 0.50 to 32 ng/mL
Assay Sensitivity 0.082 ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Recombinant MMP-2 and natural Human, Mouse, Rat, Porcine, and Canine active, pro-, and TIMP complexed MMP-2.
Sufficient For 1 x 96-well Plate
Target Species Canine, Human, Mouse, Porcine, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 4313
Gene Symbol MMP2
Quantity 1 Plate
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72 kD gelatinase, 72 kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1 matrix metalloproteinase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase)
Target MMP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.