Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Total MMP-2 Quantikine ELISA Kit
SDP

Catalog No. SMMP200 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SMMP200 1 Pk. (6 Plates)
MMP200 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SMMP200 Supplier R&D Systems™ Supplier No. SMMP200

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For human, mouse, rat, porcine, and canine samples.

The Total MMP-2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human, mouse, rat, porcine, canine MMP-2 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).

Specifications

Assay Range 0.50 to 32 ng/mL
Assay Sensitivity 0.082 ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Line R&D Systems
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum
Specificity Recombinant MMP-2 and natural Human, Mouse, Rat, Porcine, and Canine active, pro-, and TIMP complexed MMP-2.
Sufficient For 6 x 96-well Plates
Target Species Canine, Human, Mouse, Porcine, Rat
For Use With (Application) ELISA
Gene ID (Entrez) 4313
Gene Symbol MMP2
Quantity 1 Pk. (6 Plates)
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72 kD gelatinase, 72 kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP2, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1 matrix metalloproteinase 2 (gelatinase A, 72 kDa gelatinase, 72 kDa type IVcollagenase)
Target MMP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.