Learn More
R&D Systems™ Total Nitric Oxide and Nitrate/Nitrite Parameter Assay Kit
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Assay Range | 3.12 - 200 umol/L |
| Assay Sensitivity | 0.78 umol/L |
| Conjugate | Biotin |
| Cross Reactivity | < 50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Line | R&D Systems |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine |
| Specificity | Total Nitric Oxide, Nitrite and Nitrate Levels in Various Samples |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.