Learn More
ProteinSimple Total Protein Detection Module for Jess, Wes, Peggy Sue or Sally Sue
Supplier:  ProteinSimple DMTP01
Encompass
Use with a Jess/Wes or Sally Sue/Peggy Sue Separation Module. Includes: Luminol-S, Peroxide, Antibody Diluent 2, Total Protein Reconstitution Agent 2, Total Protein Streptavidin-HRP.
Catalog No. NC1197015