Learn More
Bioassay Systems QuantiChrom™ Total Protein Assay Kit. For quantitative determination of total protein.
SDP
Supplier:  Bioassay Systems QTPR100
Encompass_Preferred
For quantitative determination of total protein. Key Features: Fast and sensitive. Assay takes 10 min. Linear detection range of 5 - 200 mg/dL protein. Robust and Reliable. Substances in urine have minimal interference (< 2%). Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD600nm (Pyrogallol Red). Samples: Urine, cerebrospinal fluid etc. Species: All. Procedure: Assay takes 10 min. Kit size: 100 tests. Detection limit: 5 mg/dL.
Catalog No. 50-489-307