Learn More
Biolegend TotalSeq™-A0170 anti-human CD272 (BTLA)
Supplier:  Biolegend 344525
Encompass_Preferred
TotalSeq™-A0170 anti-human CD272 (BTLA) [MIH26]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green; Apps: PG; Size: 10 ug
Catalog No. 50-207-6665
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.