Learn More
Biolegend TotalSeq™-C0050 anti-human CD19
Supplier:  Biolegend 302265
Encompass_Preferred
TotalSeq™-C0050 anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 ug
Catalog No. NC1623475
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.