Learn More
Biolegend TotalSeq™-C0304 anti-mouse Hashtag 4
Supplier:  Biolegend 155867
Encompass_Preferred
TotalSeq™-C0304 anti-mouse Hashtag 4 [M1/42; 30-F11]; Isotype: Rat IgG2a, κ/Rat IgG2b, κ; Reactivity: Mouse; Apps: PG; Size: 10 ug
Catalog No. NC1920791
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.