Learn More
Biolegend TotalSeqTM-D0410 anti-human CD
Supplier:  Biolegend 356645
Encompass_Preferred
TotalSeq™-D0410 anti-human CD38 [HB-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: PG; Size: 10 μg
Catalog No. 50-272-2163
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.