Learn More
Whip Mix Corporation TOUCH-O-MATIC BUNSEN NAT GAS
Supplier:  Whip Mix Corporation 002807000
Encompass_Preferred
Touch-O-Matic� Bunsen Burner Natural Gas Model. Features: On-Off platform saves gas and reduces the heat load and electricity expended and also serves as a comfortable hand rest. Simply depress and turn platform slightly for continuous flame. Removable and rotatable shield protects the pilot and flame from drafts. Burner assembly can be conveniently detached for cleaning. Loop heat conductor and cup sold separately.
Catalog No. NC9891325
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.