Learn More
Thermo Scientific™ Touch Screen Dry Bath/Block Heater
Description
Designed to achieve precise temperature stability and uniformity with an easy-to-use touch screen controller. With 1, 2 and 4 block models, a broad range of heat blocks and a wide temperature range of ambient +5°C to 130°C, you're sure to find the dry bath to fit your application demands. The Thermo Scientific™ Touch Screen Dry Bath/Block Heater includes advanced features such as a timer, programmability with the ability to save up to 10 programs with 5 steps per program, and an optional external temperature sensor to control the heat setting by a real time sample read out.
- Advanced internal temperature sensing probe with outstanding temperature accuracy and control
- Temperature deviation adjustment
- Precise temperature control with PID circuit
- Temperature self-calibration enables to offset the temperature to desired value
- Timer enables the user to accurately monitor the heating time
- Password protection feature
- Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting
- Range of configuration to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications
- Powder-coated steel body construction ensures durability
- When the external temperature sensor is connected, the internal temperature sensor will disconnect automatically, allowing the temperature control and display to rely on the external temperature sensor
- Easy-to-use touch screen control displays time and temperature
- 10- user setting programs, each supports up to 5 sequential steps of different temperatures and duration times
- Built-in over temperature protection device for added safety
Applications:
- Sample preparation
- Enzyme preservation
- Enzyme-substrate reactions
- DNA amplification
- Electrophoresis gel degeneration
- Serum coagulation
Required accessory:
- Unit must be ordered with block(s)
Specifications
Specifications
| Certifications/Compliance | CE, cULus, RoHS |
| Controller Type | PID digital |
| Holds | 2 Blocks |
| Lighting | LED |
| Material | Powder-coated Steel |
| Plug Type | CN, EU, UK |
| Relative Humidity | <80% |
| Temperature Range (Metric) | Ambient +5° to 130°C (Ambient at 25°C) |
| Type | Dry Block Heater |
| Fuse | 250V 2.5A |
| Show More |
Warranty: 2-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.