Description
- Set of 20 monofilaments based on the Semmes-Weinstein monofilament set
- Features retractable filaments to protect the filament
- Allows the researcher to carry a few in a pocket
Operating Principle
When the tip of a fiber of given length and diameter is pressed against the skin at right angles, the force of application increases as long as the researcher continues to advance the probe, until the fiber bends. After the fiber bends, continued advance creates more bend, but not more force of application. This makes it possible for the researcher using a hand-held probe to apply a reproducible force, within a wide tolerance, to the skin surface.
Pre-Clinical Use
Rodents exhibit a paw withdrawal reflex when the paw is unexpectedly touched. The Touch Test™ Sensory Evaluator can be used on the Plantar surfaces of the foot of a rat or mouse. The animal indicates sensation by pulling back its paw. Individual filaments available.
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Model | Replacement Evaluator 6.10 |