Learn More
Ansell TouchNTuff™ 83-300 Ultra-Soft Polyisoprene Gloves
Catalog No. 17999807 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6.5
7.5
9
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-999-807 9
17-999-800 5.5
17-999-802 6.5
17-999-804 7.5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-999-807 Supplier Ansell Supplier No. 83300090
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Ultra-soft polyisoprene gloves offer a comfortable second skin feel for longer wear time and reduced hand fatigue.
  • Compatible with Class 100 (ISO 5) / Grade A Cleanroom Environments
  • Latex-free polyisoprene material offers the performance of natural rubber latex without the risk of latex sensitization
  • Ideal for double donning
  • Prevents Type I allergies
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577, ASTM D7160
Certifications/Compliance (Europe) CE 0086, Category III, EN 420:2003 + A1:2009, EN 455 1-4
Color White
Latex Free Yes
Material Polyisoprene
Size 9
EN 421 Yes
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES Type B
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Beaded
Manufacturing/QMS Audit Standards ISO 10282, ISO 13485, ISO 14001, ISO 9001
Model 83300090
Series TouchNTuff™ 83-300
Length (English) 11.6 in.
Length (Metric) 29.5 cm