Learn More
Ansell TouchNTuff™ 83-500 Sterile Polyisoprene Gloves
GREENER_CHOICE
Catalog No. 17000839 Shop All Ansell Products
Change view
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-839 8
17-000-834 5.5
17-000-837 7
17-000-835 6
17-000-836 6.5
17-000-838 7.5
17-000-840 8.5
17-000-841 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 17-000-839 Supplier Ansell Supplier No. 83500080
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterile polyisoprene gloves are ergonomically-designed to enhance fit and comfort, and reduce hand fatigue and stress.
  • Powder-free, chlorinated glove interior
  • Rolled bead cuff style
  • Hand specific
  • Smooth finish
  • Polyisoprene
  • White
TRUSTED_SUSTAINABILITY
Specifications
Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 0.65
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Polyisoprene
Powdered Powder Free
Product Type Cleanroom Gloves
Size 8
Sterility Sterile
Thickness (Metric) 0.21 mm
Glove Cuff Style Beaded/Rolled
Model 835008
Quantity 200/Pk.
Resistant To Splash resistance
Series 83-500
Length (English) 12
Length (Metric) 300
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.