Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves PROMO

Catalog No. 1892748DW25 Shop All Ansell Products
Change view
Click to view available options
Ansell TouchNTuff™ Disposable Nitrile Gloves
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-927-48DW25 Ansell TouchNTuff™ Disposable Nitrile Gloves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-927-48DW25 Supplier Ansell Supplier No. 1892748DW25

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

  • Made from 100% nitrile
  • Resistant to nuisance punctures, nicks, cuts and abrasions
  • Comfortable thin mil construction
  • Rolled cuffs for easy donning and doffing
  • Chemical resistant
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671
Certifications/Compliance (Europe) Category III, EN 420:2003 + A1:2009
Chlorinated No
Color Green
Disposable Single-use
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Size X-Large
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
EN 421 No
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES JKPT
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Beaded/Rolled
Lining Unlined
Manufacturing/QMS Audit Standards ISO 9001
Model 92 600 XL585837
Series TouchNTuff 92-600
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.