Learn More
Brady™ ToughBond™ B-483 Gloss Polyester Labels with an Aggressive Adhesive
Shop All Brady Corporation ProductsSpecifications
Specifications
| Certifications/Compliance | UL Recognized, CSA Approved |
| Description | Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant |
| Adhesive Type | Rubber, Permanent |
| Color | White |
| Finish | Matte |
| Material | Polyester (B-483) |
| Product Type | Asset Tracking Label |
| For Use With (Application) | Component ID, Equipment ID, Barcoding, Panel ID, Voice/Data ID, Asset tracking, Rating plates, General purpose ID |
| For Use With (Equipment) | BMP™71 Printer |
| Quantity | 1 Ea. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.