Learn More
Brady™ ToughBond™ B-489 Matte Polyester Labels with an Aggressive Adhesive
Shop All Brady Corporation ProductsSpecifications
Specifications
| Certifications/Compliance | UL Recognized, CSA Approved |
| Description | Chemical, Corrosion, Humidity, UV and Weather-Resistant |
| Adhesive Type | Rubber, Permanent |
| Color | White |
| Finish | Matte |
| Material | Polyester (B-489) |
| Product Type | Asset Tracking Label |
| For Use With (Application) | Asset Tracking; Barcode Labeling; Component and Equipment ID; General and Industrial ID; Panel Labeling; Rating Plates; Voice and Data ID |
| For Use With (Equipment) | BMP™71 Printer |
| Labels per Row | 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.