Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ToughStripe™ Anti-Slip Floor Safety Sign

Catalog No. 19150823 Shop All Brady Corporation Products
Change view
Click to view available options
Displayed Text:
BIOHAZARD
Chemical Storage
Ear Protection Area
Emergency Exit
Exit
Eye Protection Area
Eye Wash Station
Fire Alarm
Fire Extinguisher
Fire Hose
First Aid
Hard Hat Area
Safety Glasses Required
Safety Goggles Required
Safety Shower
Watch Your Step
Color:
Black
Black on Orange
Black on White
Black on Yellow
Red
White
Shape:
Rectangular
Round
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text Color Shape
19-150-823 Emergency Exit Black on White Round
19-150-815 Safety Shower Black Round
19-150-814 First Aid White Round
19-150-818 Exit Black on White Round
19-150-819 First Aid White Rectangular
19-150-820 Fire Extinguisher Black on White Round
19-150-826 Chemical Storage Red Rectangular
19-150-827 Watch Your Step Black on Yellow Rectangular
19-150-830 Ear Protection Area Black on Yellow Rectangular
19-150-831 Eye Protection Area Black on Yellow Rectangular
19-150-832 Hard Hat Area Black on Yellow Rectangular
19-150-835 Eye Wash Station Black Round
19-150-838 Safety Glasses Required Black on Yellow Rectangular
19-150-822 Fire Hose Black on White Round
19-150-821 Fire Alarm Black on White Round
19-150-834 BIOHAZARD Black on Orange Rectangular
19-150-833 Safety Goggles Required Black on Yellow Rectangular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 19-150-823 Supplier Brady™ Supplier No. 104495
Add to Cart
Add to Cart

Hard-to-miss signs made of durable floor marking tape

  • Backed with high-performance adhesive
  • Features a highly visible, glossy appearance for communicating important safety and warning messages to employees
  • Clear anti-skid area
  • Designed with sections of anti-skid material that help prevent accidental slips, trips and falls
  • Easy to remove and apply
  • Made of polyester

Specifications

Certifications/Compliance ASTM ANSI F1679, ANSI A1264.2-2006, OSHA
Alternate Part Number B534FS 17 in. RD,BK/WT EMERGENCY EXIT W/SYM
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Rubber
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
For Use With (Application) Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements with glossy appearance and clear anti-skid area for communicating important safety and warning messages to employees
Max. Temperature (English) 129.9°F
Min. Temperature (English) -0.4°F
Thickness 0.01 in.
Temperature Range (Metric) -18°C to 54°C
Shape Round
Temperature Range (English) -0.4°F to 129.9 °F
Languages English
Displayed Text Emergency Exit
Product Type Floor Marking Safety Sign
Color Black on White
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.