Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ToughStripe™ Premium 5S Marking Kit with BMP™71 Label Printer

Catalog No. 19104536 Shop All Brady Corporation Products
Change view
Click to view available options
Packaging Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-104-536 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-536 Supplier Brady™ Supplier No. 142111

This ToughStripe™ Premium 5S Marking Kit provides everything you need to 5S your facility, including a BMP™71 Portable Label Printer and supplies for creating custom labels.

  • Kit includes ToughStripe™ floor marking tapes, tool board, and workbench marking tapes compatible with the BMP™71 Label Printer.
  • ToughStripe™ floor marking tapes are made of rugged polyester with ultra-aggressive adhesive to withstand forklift traffic without tearing or lifting
  • The painless setup, portability and intuitive interface of the BMP™71 label printer makes it easy to create custom labels on the go
  • 5S lean methodology reduces waste and optimizes productivity through better workplace organization

Specifications

Certifications/Compliance FCC Part 15 Class A
Displayed Text Blank
Height (English) 13 in.
Width (English) 9.5 in.
Packaging Quantity 1 Kit
Type Marking Tape
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.