Learn More
ATCC TOV-112D (CRL-3593) human ovarian carcinoma epithelial cell line - frozen vial
Supplier:  ATCC CRL3593
Encompass
TOV-112D is a human epithelial ovarian carcinoma cell line derived in 1992 from the ovary of a 42-year-old female with primary malignant adenocarcinoma. Supplied frozen for research use, the cell line is adherent, tumorigenic, and used in ovarian cancer studies.
  • Epithelial morphology.
  • Adherent growth properties.
  • Tumorigenic in nude mice; forms colonies in soft agar.
  • Her2/neu positive; p53 Arg→His mutation (exon 6, codon 175).
  • Karyotype: 52, XX, add(X)(p22), +add(1)(p22), +add(1)(p22), +2, +9, +12, add(15)(p11), +17.
  • Recommended medium: 1:1 MCDB-105 and Medium 199 supplemented with 15% fetal bovine serum.
  • Product supplied frozen; store in vapor phase of liquid nitrogen (≤ -130 °C).
  • Mycoplasma not detected; STR profile available.
Catalog No. NC3358822