Learn More
Med Vet International OR Towels, 16 x 26-in Blue, Each
Supplier:  Med Vet International AHSORTB1
Encompass_Preferred
New 16" x 26" 100% cotton towels. These blue surgical towels are reusable and can be autoclaved. Long-lasting, top-quality and durable. Highly absorbent, non-sterile.
Catalog No. 50-252-2455
Due to product restrictions, please to purchase or view availability for this product.