Description
- Get the complete safety package – unlike signs and tapes this cover provides a sign and an unavoidable boundary in one easy-to-use product
- Perfect for manways that need to be locked out or secured, but little or no flange is present
- Lock it down – get another level of safety with lockable options that give authorized personnel the option to apply a padlock to maintain restrictive access
- Includes (2) mesh covers and (2) ratchet straps
Specifications
Specifications
| Product Type | Confined Space Cover |
| Color | Black/Red |
| Material | Metal |
| Dimensions (L x W) | 40 x 40 in. |
| Displayed Text | Confined space. Enter by permit only. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.