Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Triage TOX Drug Screen Evaluation Panel
SDP

Catalog No. 1398232
Change view
Click to view available options
Quantity: 20 x 3.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-982-32 Quantity: 20 x 3.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-982-32 Supplier Quidel Triage Supplier No. 26657

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel Triage TOX Drug Screen Evaluation Panel is designed for toxicology validation applications.

  • Used for method validation with Quidel Triage TOX Drug Screen 1398229 and Quidel Triage Rapid Visual Read tests

Specifications

For Use With (Application) Method validation with TOX Drug Screen 1398229 and Rapid Visual Read tests
Storage Requirements Frozen (-20°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.