Learn More
Drg International Inc DRG Toxoplasma Gondii IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3519R
Encompass_Preferred
An enzyme immunoassay for the quantitative and qualitative determination of IgG-class antibodies to Toxoplasma gondii in human serum and plasma. The DRG Toxoplasma gondii IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) for Research Use Only. Microtiter wells, as a solid phase, are coated with inactivated Toxoplasma gondii soluble antigen (strain RH). Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.