Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Toxoplasma gondii Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA173002
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173002 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173002 Supplier Invitrogen™ Supplier No. PA173002
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The PA1-73002 antibody reacts with Toxoplasma gondii. PA1-73002 has been successfully used in ELISA applications. The PA1-73002 antibody was raised against the RH Strain of T. Gondii.

Toxoplasma is a crescent shaped sporozoan that lives as an intracellular parasite in various tissues of vertebrates and completes its life cycle in a single host. It is transmitted via raw/undercooked meat, contaminated soil, or by direct contact. Infection due to Toxoplasma gondii usually occurs in pregnant women where a variable degree of immunosuppression may exist or in patients receiving immunosuppressive drug therapy following organ transplant. Toxoplasma infects tissue of the GI tract where an active infection is accompanied by fever and enlargement of the spleen. Symptoms of toxoplasmosis are generally mild but severe infection of lymph nodes may occur. Congenital toxoplasmosis, in which the maternal infection is transmitted during pregnancy, can produce blindness or mental retardation in the newborn.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Toxoplasma gondii
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias t gondii; T. gondii
Host Species Goat
Immunogen RH Strain of T. Gondii.
Purification Method IgG Fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.