Learn More
Description
This antibody recognizes the SAG1 protein of T. gondii. Purity is >90%.
Specifications
Specifications
| Antigen | Toxoplasma gondii SAG1 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D61S |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | t gondii; T. gondii; T. gondii SAG1 |
| Host Species | Mouse |
| Immunogen | Toxoplasma gondii native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
