Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOYOPEARL™ LABPAK Bulk Media Sampler, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022216 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2216 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2216 Supplier MilliporeSigma Supelco Supplier No. 843310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Allow the use of several resins to determine which works best for a particular application

  • LabPak products are multi-milliliter containers of TOYOPEARL and TSKgel bulk media products.
  • Typically they contain 3 or 4 different ligand types offered for a particular chromatography mode.
  • The larger resin amounts in LabPak products allow the packing of wider ID and longer columns than available in the ToyoScreen products.
  • Allow the use of several TOYOPEARL™ resins to determine which works best for a particular application. TOYOPEARL™ resins are used in separation media and resins.
TRUSTED_SUSTAINABILITY

Specifications

For Use With HPLC: Suitable
Particle Size 45 μm
Quantity 3/Pk.
Volume (Metric) 150 mL

Toyopearl is a registered trademark of Tosoh Corporation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.