Learn More
Invitrogen™ TP53INP2 Polyclonal Antibody

Description
Immunogen has 81% identity to rat. Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended.
Specifications
Specifications
| Antigen | TP53INP2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.05% sodium azide |
| Gene | TP53INP2 |
| Gene Accession No. | Q8CFU8, Q8IXH6 |
| Gene Alias | 1110029F20Rik; C20orf110; diabetes and obesity regulated; diabetes and obesity-regulated; Diabetes and obesity-regulated gene; Diabetes and obesity-regulated protein; dJ1181N3.1; DOR; p53-inducible protein U; PIG-U; PINH; TP53INP2; transformation related protein 53 inducible nuclear protein 2; Trp53inp2; tumor protein p53 inducible nuclear protein 2; tumor protein p53-inducible nuclear protein 2 |
| Gene Symbols | TP53INP2, Trp53inp2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.