Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TP53INP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575928 Supplier Invitrogen™ Supplier No. PA575928
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

TP53INP2 (tumor protein p53 inducible nuclear protein 2), also known as DOR or PINH, is a 220 amino acid nuclear protein that is expressed in developing murine brain and spinal cord, as well as in the sensory and motor neuron tracts of the peripheral nervous system. A novel protein involved in the autophagy of mammalian cells, TP53INP2 translocates from the nucleus to the autophagosome structures after activation of autophagy by rapamycin or starvation. Necessary for autophagosome development and considered a scaffold protein, TP53INP2 recruits LC3 and/or LC3-related proteins, such as GABARAP and GABARAP-like2, to the autophagosome membrane by interacting with the transmembrane protein TMEM49. The gene encoding TP53INP2 is located on human chromosome 20, which is comprised of approximately 2% of the human genome and contains nearly 63 million bases that encode over 600 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TP53INP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene TP53INP2
Gene Accession No. Q8CFU8, Q8CHM3, Q8IXH6
Gene Alias 1110029F20Rik; C20orf110; diabetes and obesity regulated; diabetes and obesity-regulated; Diabetes and obesity-regulated gene; Diabetes and obesity-regulated protein; dJ1181N3.1; DOR; p53-inducible protein U; PIG-U; PINH; TP53INP2; transformation related protein 53 inducible nuclear protein 2; Trp53inp2; tumor protein p53 inducible nuclear protein 2; tumor protein p53-inducible nuclear protein 2
Gene Symbols TP53INP2, Trp53inp2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 148-193 of Human TP53INP2.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362246, 58476, 68728
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.