Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TP63 Rabbit anti-Human, Mouse, Rat, Unconjugated, Clone: TP63/1423R, Abnova™

Catalog No. 89339742 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-742 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-742 Supplier Abnova Corporation Supplier No. RAB00703
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

TP63 Monoclonal antibody specifically detects TP63 in Human, Mouse, Rat samples. It is validated for IHC-P.

Specifications

Antigen TP63
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TP63/1423R
Conjugate Unconjugated
Description tumor protein p63
Gene TP63
Gene Alias AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP73L, p40, p51, p53CP, p63, p73H, p73L
Host Species Rabbit
Immunogen Recombinant human p63 protein fragment
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8626
Target Species Human, Mouse, Rat
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.