Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TPA Antibody, ProSci
SDP

Catalog No. 16590878
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-590-878 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-590-878 Supplier ProSci LLC Supplier No. 624570400
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TPA Polyclonal Antibody specifically detects TRAP in Human Samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry.

Specifications

Antigen TPA
Applications Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry
Classification Polyclonal
Conjugate Unconjugated
Dilution For WB starting dilution is: 1:1000, For IHC-P starting dilution is: 1:10~50, For FACS starting dilution is: 1:10~50
Formulation Supplied in PBS with 0.09% (W/V) sodium azide.
Gene Accession No. P00750
Gene Alias Tissue-type plasminogen activator, t-PA, t-plasminogen activator, tPA, Alteplase, Reteplase, Tissue-type plasminogen activator chain A, Tissue-type plasminogen activator chain B, PLAT
Host Species Rabbit
Immunogen This TPA antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 371-399 amino acids from the Central region of human TPA.
Purification Method This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
Quantity 0.4 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5327
Target Species Human
Content And Storage Store at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Form Liquid
Isotype Ig
Research Category Cancer, Cell Cycle, Immunology, Obesity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.