Learn More
Haematologic Technologies Inc TPA SUBSTRATE 1MG
Supplier:  Haematologic Technologies Inc SN18
Encompass_Preferred
Fluorogenic substrate, Contains the fluorescent reporter group 6-amino-1-naphthalene-sulfonamide (ANSN) for monitoring enzyme activity of tPA,Boc-L-(p-F)FPR-ANSNH-C2H5, 1 milligram, Research use only, Supplied in DMSO, store at -20 Celsius.
Catalog No. 50-883-411
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.