Learn More
Medchemexpress LLC HY-131614 5mg Medchemexpress, TPC2-A1-N CAS:136186-07-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1316145MG
Encompass_Preferred
Medchemexpress, HY-131614 5mg TPC2-A1-N CAS:136186-07-7 TPC2-A1-N is a powerful and Ca2+-permeable agonist of two pore channel 2 (TPC2), which plays its role by mimicking the physiological actions of NAADP. TPC2-A1-P reproducibly evokes significant Ca2+ responses from TPC2 (EC50=7.8 μM), and the effect can be blocked by several TPC blockers. TPC2-A1-N can be used to probe different functions of TPC2 channels in intact cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1288
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.