Learn More
Abcam TPCA 1, IκB Kinase Inhibitor, >98% Purity, Solid, 10 mg
Description
- Molecular Weight: 279.29 Da
- Purity: >98%
- Potency: Potent, selective IκB kinase inhibitor (IC50 = 17.9 nM)
- Mechanism: Inhibits NF-κB nuclear localization
- In vivo effects: Shows anti-inflammatory effects
- Form: Solid
- Storage: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | IκB Kinase |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% Tpca-1 |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% Tpca-1 |
| Quantity | 10 mg |
| For Use With (Application) | IkappaB kinase inhibitor, NF-kB nuclear localization inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.