Learn More
Worthington Biochemical Corporation TPCK TREAT.TRYPSIN.22UM 5X50MG
SDP
Supplier:  Worthington Biochemical Corporation LS003734
Encompass_Preferred
Trypsin chromatographically purified, diafiltered (Code TRL3) filtered thorugh a 0.22 micron pore size membrane and lyophilized in sterile vials. This product is not tested for pyrogenicity.
Catalog No. NC9545485
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.