Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tpd52 (Tumor Protein D52) Polyclonal Antibody, Abbomax

Catalog No. 502203912
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3912 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3912 Supplier Abbomax Inc Supplier No. 600610
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF-7 cells Cellular location: Cytoplasmic, Membrane.

Tpd52 (Tumor Protein D52) Polyclonal antibody specifically detects Tpd52 (Tumor Protein D52) in Bovine, Chicken, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Tpd52 (Tumor Protein D52)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Tpd52
Gene Accession No. P55327, Q62393
Gene Alias 28 kDa calcium-dependent phosphoprotein; AI043038; calcium sensitive phosphoprotein (CSPP28); CSPP28; D52; hD52; LOC100058422; mD52; N8L; PC-1; pp28; PrLZ; prostate and colon associated protein; prostate leucine zipper; protein N8; si:ch211-203l9.2; si:ch211-203l9.3; TPD52; tumor promoter D52 protein; tumor protein D52; Unknown (protein for MGC:127812)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the internal sequence of tumor protein D52 (Tpd52)
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21985, 294900, 540235, 7163, 770339
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.