Learn More
TPH in Water (High Level), Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017021 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1.2 mL
Catalog No. Quantity
11-101-7021 1.2 mL
1 options
Catalog No. 11-101-7021 Supplier MilliporeSigma Supelco Supplier No. PE16191.2ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This vial contains a concentrate of a blended gasoline from three different sources and diesel fuel oil No. 2 in pentane for analysis by TNRCC Method 1005. When prepared according to instructions, the concentration of each analyte will be less than 100 mg/L. The packsize on this sample has recently changed from 2ML to 1.2ML. This change does not affect the performance of the proficiency test.Sample preparationTo prepare solution, fill a 40-mL voa vial to volume with carbon-filtered reagent-grade water at room temperature. Remove enough water to allow for the addition of extraction solvent. Open vial PE1619-1.2ML and immediately withdraw 40 muL with a gas tight syringe. Quickly inject the 40 muL aliquot below the surface of the reagent water in the voa vial. Add the extraction fluid. Mix the sample by inverting the flask three times only (excessive shaking results in loss of volatiles) and analyze immediately. Assume a 40mL sample for calculation purposes. Note this is a 1:1000 dilution and other dilutions are acceptable.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 1.2 mL
Concentration or Composition (by Analyte or Components) Residual Range Organics (RRO),>C28-C35 1-100 mg/L; Total Petroleum Hydrocarbons (TPH), (C6-C35 20-300 mg/L; Diesel-range total petroleum hydrocarbons, >C12-C28 1-100 mg/L; Gasoline Range Organics, C6-C12 1-100 mg/L)