Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPH2 Monoclonal Antibody (CL2990)
GREENER_CHOICE

Catalog No. PIMA535008 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535008 Supplier Invitrogen™ Supplier No. MA535008
  • $630.00
    $510.65 / Each of 1

    Save $119.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Immunogen sequence: SITRPFSVYF NPYTQSIEIL KDTRSIENVV QDLRSDLNTV CDALNKMNQY LG Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

Tryptophan hydroxylase (TPH) is a member of the pterin-dependent aromatic acid hydroxylase family. It catalyzes the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. TPH is expressed predominantly in the brain stem. Mutations in this gene may be associated with psychiatric diseases such as bipolar affective disorder and major depression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPH2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2990
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Tph2
Gene Accession No. Q8CGU9, Q8CGV2, Q8IWU9
Gene Alias ADHD7; AU043594; MGC119994; Neuronal tryptophan hydroxylase; NTPH; Tph2; TPRH; TRPH; Tryptophan 5-hydroxylase 2; tryptophan 5-monooxygenase 2; tryptophan hydroxylase 2
Gene Symbols Tph2
Host Species Mouse
Immunogen Recombinant protein fragment of Human TPH2 (Product #RP-98454)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121278, 216343, 317675
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.