Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TPM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529846 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529846 Supplier Invitrogen™ Supplier No. PA529846
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse liver, rat muscle. Predicted reactivity: Mouse (99%), Rat (88%), Zebrafish (80%), Xenopus laevis (83%), Pig (88%), Rabbit (88%), Chicken (81%), Sheep (88%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Tropomyosin is a dimeric coiled-coil protein that binds along the length of actin filaments. It is associated with the thin filaments of muscle cells and the microfilaments of nonmuscle cells. Chicken embryo fibroblasts (CEF) contain five isoforms of tropomyosin (a, b, 1, 2, and 3), identified as such by their different apparent molecular masses after separation by SDS-PAGE, but similar biochemical properties, such as resistance to heat and organic solvents, the ability to bind to F-actin filaments, and the lack of proline and tryptophan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TPM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene TPM1
Gene Accession No. P04692, P09493, P42639, P58771
Gene Alias AA986836; AI854628; alpha tropomyosin; alpha tropomyosin of brain; alpha tropomyosin of fibroblasts F2; alpha-fTM; Alpha-tm; alpha-Tm-1; alpha-Tm-2 isoform; alpha-tropomyosin; alpha-tropomyosin (partial); alpha-tropomyosin 1; alpha-tropomyosin 2; alpha-tropomyosin of skeletal fast muscle; C15orf13; cardiac alpha tropomyosin; cardiomyopathy, hypertrophic 3; CMD1Y; CMH3; epididymis secretory protein Li 265; HEL-S-265; hepatoma alpha tropomyosin; HTM-alpha; LVNC9; sarcomeric tropomyosin kappa; smooth muscle alpha-tropomyosin; striated muscle alpha-tropomyosin; Tm1; TM2; Tm3; Tma2; Tmpa; Tmsa; Tpm1; Tpm-1; tpm1 protein; TPM1-1; TPM1-alpha; TPMA; tropomyosin; tropomyosin (CTm4); tropomyosin (CTm7); tropomyosin 1; tropomyosin 1 (alpha); tropomyosin 1 alpha chain; tropomyosin 1, alpha; tropomyosin 1-1; tropomyosin 3 alpha; tropomyosin alpha-1 chain; Tropomyosin-1; unnamed protein product; zgc:171719; zgc:55951; zgc:77009
Gene Symbols TPM1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Tropomyosin 1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100037999, 22003, 24851, 7168
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.