missing translation for 'onlineSavingsMsg'
Learn More

TPM3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 554441AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-6163


May include imposed supplier surcharges.
Only null left
Add to cart

Description

Description

TPM3 (tropomyosin 3) is a member of the tropomyosin family of actin-binding proteins involved in the contractile system of striated and smooth muscles and the cytoskeleton of non-muscle cells. Tropomyosins are dimers of coiled-coil proteins that polymerize end-to-end along the major groove in most actin filaments. They provide stability to the filaments and regulate access of other actin-binding proteins. In muscle cells, they regulate muscle contraction by controlling the binding of myosin heads to the actin filament. Mutations in this gene result in autosomal domit nemaline myopathy, and oncogenes formed by chromosomal translocations involving this locus are associated with cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

TPM3
Polyclonal
Unconjugated
TPM3
Gamma tropomyosin, hscp30, hTM5, NEM1, OK/SW cl.5, TM 5, TM3, TM30, TM30nm, TPM3, TPMsk3, TRK, tropomyosin 3, Tropomyosin 5, Tropomyosin alpha 3 chain
Rabbit
Antigen Affinity Chromatography
RUO
59069, 7170
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P06753, P21107
Tpm3
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.