Learn More
Sigma Aldrich Fine Chemicals Biosciences TPPR easy read cell counte1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z761001
Encompass
Features of the easy read mesuring device eading device with magnifying glas and cross lineoptimal usage in combination with the PCV tubereproducible cell pellet measuring possiblereusable product no further investments necessarymeasuring range 0 - 5 mul
Catalog No. 50-290-8552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.