Learn More
Epredia™ TPX Single Sample Chamber, Caps and Filter Cards
Catalog No. 5991022 Shop All Epredia Products
Change view
Click to view available options
Volume (Metric):
0.1 to 0.4 mL
0.1 to 0.5 mL
Type:
Filter Card
TPX Sample Chamber
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Type
5991022 0.1 to 0.5 mL Filter Card
A78710018 0.1 to 0.5 mL TPX Sample Chamber
5991023 0.1 to 0.4 mL Filter Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 5991022 Supplier Epredia™ Supplier No. 5991022
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with disposable white filter cards for sample volumes up to 0.5 mL and for use with disposable brown filter cards for sample volumes of 0.4 mL or less.
TPX Single Sample Chamber
  • Reusable
  • Autoclavable
Filter Cards
  • Highly absorbent disposable filter cards for use with TPX chamber
  • Not for use with Cytofunnel™ disposable sample chambers
Specifications
Color White
Description White Filter Cards
Disposable Single-use
Type Filter Card
Volume (Metric) 0.1 to 0.5 mL