Learn More
TARGETMOL CHEMICALS INC TR-14035 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T53105MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.TR-14035 (MDK-1191) is a dual antagonist of alpha4beta7/alpha4beta1 integrins (IC50s 7/87nM). Purity 99.81%
Catalog No. 50-287-6790
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.