Learn More
OTT Hydromet Adcon - TR2 - SDI 12
Supplier: OTT Hydromet 200.733.032
The temperature sensor offers an accuracy of 0.05°C. Including all potential sources of errors, repeatability, hysteresis, linearity, etc., its accuracy is still better than 0.1°C. The temperature measurement range was extended to +80°C. The humidity sensor is almost fully encapsulated to provide maximum protection against particles. The measurement window is specially coated to reduced all possible negative effects of dust, moisture and (agro-)chemicals. The sensor covers the full range up to 100%, with a typical accuracy of 1% up to 90% and 2% from 90 to 100%. TR2 also calculates dew point directly inside the sensor, and outputs even the mixing ratio. A highly professional 7-element radiation shield protects the sensor from direct sunlight and provides sufficient ventilation. The delivery pack contains the sensor, a 7-element radiation shield, mast mounting gear for poles with a diameter of ~40mm and a 3m cable with a 7-pin Binder connector for Adcon RTUs.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.