Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRA-1-60 Monoclonal Antibody (cl.A)
GREENER_CHOICE

Catalog No. 411000
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-100-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-100-0 Supplier Invitrogen™ Supplier No. 411000
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

TRA-1-60 Monoclonal Antibody for Western Blot, ICC/IF

TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-60
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone cl.A
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias TRA160; tra60
Host Species Mouse
Immunogen 2102Ep cl.2A6 human embryonal carcinoma cells.
Purification Method Affinity chromatography - MBP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.