Learn More
Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60), DyLight™ 550

Description
MA1-023-D550 has been successfully used in ICC/IF applications on human samples.
Specifications
Specifications
| Antigen | TRA-1-60 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | TRA-1-60 |
| Concentration | 1 mg/mL |
| Conjugate | DyLight 550 |
| Formulation | PBS with proprietary stabilizer and 0.02% sodium azide |
| Gene Alias | TRA160; tra60 |
| Host Species | Mouse |
| Immunogen | Human embryonal carcinoma cell line 2102Ep. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.