Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRA-1-60 Monoclonal Antibody (TRA-1-60), DyLight™ 650
GREENER_CHOICE

Catalog No. MA1023D650 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA1023D650 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA1023D650 Supplier Invitrogen™ Supplier No. MA1023D650
  • $537.00
    $408.65 / Each of 1

    Save $128.35

    24% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-023-D650 has been successfully used in ICC/IF and flow cytometry applications on human samples.

TRA-1-60 is a cell surface antigen, expressed along with SSEA-3, SSEA-4 and TRA-1-81 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are lost during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRA-1-60
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone TRA-1-60
Concentration 1 mg/mL
Conjugate DyLight 650
Formulation PBS with proprietary stabilizer and 0.02% sodium azide
Gene Alias TRA160; tra60
Host Species Mouse
Immunogen Human embryonal carcinoma cell line 2102Ep.
Purification Method Affinity chromatography - MBP
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.